Anti-PRLR Rabbit Polyclonal Antibody

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILP2103-C01 ILP2103-C05 ILP2103-C1
ILP2103-C01EA 150 EUR
ILP2103-C01 ILP2103-C05 VWRKILP2103-C1
Anti-PRLR Rabbit Polyclonal Antibody
Antilichamen
Anti-Prolactin is a useful marker in classification of pituitary tumors and the study of pituitary disease. It reacts with prolactin-producing cells. Such prolactin-producing cells can also be found in prostate epithelium.

  • Visualization: cytoplasmic
  • Control: pituitary
  • Dilution Range: 1:100-1:500

Type: Primary
Antigen: PRLR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR