Syringes for gas chromatography autosamplers, CTC/LEAP PAL GC, S-Line
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
67436-01 67430-01 67434-01 67438-01 67440-01 67454-01
HAMI67436-01EA 127 EUR
HAMI67436-01 HAMI67430-01 HAMI67434-01 HAMI67438-01 HAMI67440-01 HAMI67454-01
Syringes for gas chromatography autosamplers, CTC/LEAP PAL GC, S-Line
Spuiten Injectiespuiten voor chromatografie
S-Line syringes are the cost-effective choice for CTC PAL autosampler applications. The Gastight® syringes have a precision-machined PTFE plunger tip which creates a leak-free seal that is an ideal choice for HPLC sample injections. Microliter syringes incorporate a hand-fitted stainless steel plunger with a finely bored syringe barrel.