ESD safety shoes, lace-up, New Classic, Leroy S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
726281/38 726281/39 726281/40 726281/41 726281/42 726281/46 726281/47 726281/48
ELTN726281/38EA 78.9 EUR
ELTN726281/38 ELTN726281/39 ELTN726281/40 ELTN726281/41 ELTN726281/42 ELTN726281/46 ELTN726281/47 ELTN726281/48
ESD safety shoes, lace-up, New Classic, Leroy S3
Schoenen en laarzen
ESD safety shoes made from cowhide leather, a full padded bellows tongue plus a dual-density PU/PU sole (New Classics).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), black
  • Soft fleece ESD insole
  • Scuff cap protects the leather upper
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificaten: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.