VWR®, Flessenassemblages, PETG, steriel, voor eenmalig gebruik
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
734-2844EA 940 EUR
734-2844 734-2845 734-2846 734-2850 734-2851 734-2894
VWR®, Flessenassemblages, PETG, steriel, voor eenmalig gebruik
Flessen Mediumflessen
Sterile PETG containers, with weldable tubings, provide the maximum amount of flexibility and reliability. These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. They come in a comprehensive size selection with tubing that is completely weldable/sealable.
  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • When used with sterile VWR® tube sets, custom possibilities are endless
  • Gamma irradiated to Sterility Assurance Level (SAL) 10⁻⁶
Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, also production and cleanroom applications. All product contact materials are animal-free or processed in accordance to EMEA/410/01. The assemblies are able to withstand temperatures from –20 to +65 °C without losing integrity. All assemblies are individually double-bagged with product label located on the inner bag. Gamma irradiated to Sterility Assurance Level (SAL) 10-6.
Opgelet: *The product(s) on this page are outside the scope of the EU Directive 2019/904.