Veiligheidsspuitflessen, smalle hals, Right-to-Understand, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2429-0501 2429-0502 2429-0503 2429-0504 2429-0505 2429-0506
215-8100EA 322 EUR
215-8100 215-8109 215-8110 215-8119 215-8120 215-8125
Veiligheidsspuitflessen, smalle hals, Right-to-Understand, Nalgene®
Flessen Spuitflessen
Narrow, cylindrical FEP bottle with ETFE dispensing cap. New globally harmonised chemical labeling rules have been enacted to protect workers who come into contact with hazardous chemicals.
  • Greater awareness of hazards, resulting in safer use of chemicals in the workplace; detailed safety information at your fingertips
  • 100% GHS information (no mixing of systems): wash bottles are labelled with the same warning system as is used on purchased solvents
  • Chemically inert (low leachable) making it suitable for storing high purity solvent
  • Autoclavable, with stem and draw tube
  • Bottles are not vented
De flessen zijn vooraf gelabeld met chemische identificatie en aanwijzingen voor veilig gebruik, volgens de specificaties van het GHS die van toepassing zijn op secundaire labocontainers. De spuitflessen zijn geschikt voor de overdracht van agressieve laboratoriumchemicaliën vanuit containers van de fabrikant.
De veiligheidsinformatie van het solvent is permanent gelaserd, waardoor de informatie nooit losraakt of onleesbaar kan worden doorheen de levensduur van de fles (FEP-spuitflessen zijn bekend om moeilijk te labelen door het gladde oppervlak. Markeerstift, waskrijt en labels blijven niet plakken).
Bottles are made from high-purity FEP for low extractables and long life in the lab, and are suitable for dispensing high-purity solvents. Permanently laser etched with 100% Globally Harmonized System (GHS) chemical safety information. No other system markings are used on the bottles, which eliminates confusion. Safety information is laser etched directly into the wall of the bottle so information will remain permanent and legible for the life of the wash bottle. Dispensing nozzles are carefully molded to deliver a straight stream for precision dispensing and safety. Designed to dispense through the top with a closure and stem molded in one piece for uniform, leakproof service.
Nalgene Right-to-Understand FEP wash bottles are ready to use when transfering common laboratory chemicals from manufacturers’ containers. They are pre-labeled with chemical identification information, hazard and precautionary statements, GHS pictograms, CAS, and EC numbers. Wash bottles are available for use with acetone, toluene, n-hexane, methylene chloride, xylene, and methyl ethyl ketone (MEK).
Certificaten: Labeled with Globally Harmonized System (GHS) chemical information.