Cyclic-GMP fluorescent assay kits, CatchPoint™
Leverancier: Molecular Devices
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R8065 R8074 R8075
MLDVR8065EA 2460 EUR
MLDVR8065 MLDVR8074 MLDVR8075
Cyclic-GMP fluorescent assay kits, CatchPoint™
Testen Cellulaire assays
CatchPoint™ cGMP Fluorescent Assay Kits measure cGMP levels via a competitive immunoassay format. The second messenger cyclic GMP (cGMP) modulates cellular processes, including smooth muscle relaxation, kidney function, and inflammation, by activating effectors such as protein kinases, cGMP-dependent phosphodiesterases, and cGMP-gated ion channels.
  • High affinity reagents are optimised for sensitivity when cGMP levels are low, ensuring results when other assays fail
  • Robust format resists interference from colored or fluorescent compounds, improving reliability of results
  • Rapid signal development (<10 minutes) and no termination step give users a flexible read time that is compatible with automation and high-throughput screening
  • Quantitative method allows accurate determination of cGMP in samples
The high-affinity reagents in the CatchPoint® cGMP Fluorescent Assay Kits are optimised for sensitivity and precision in applications where cGMP levels are low. A single wash step removes unbound material prior to the development step, so the assay is very resistant to interference from coloured or fluorescent test compounds. The proprietary Stoplight Red substrate is used to generate a stable fluorescent readout, allowing the assay to be read in as little as 10 minutes or up to 24 hours after substrate addition.
Leveringsinformatie: All kits contain microplates, ELISA Assay Buffer,l Rabbit Anti-cGMP Antibody, cGMP Calibrator, HRP-cGMP Conjugate, 10X Wash Solution Concentrate, Cell Lysis Buffer, 100X Substrate, and Substrate Buffer.