Potassium assay kits, FLIPR®
Leverancier: Molecular Devices
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R8222 R8223 R8330
MLDVR8222EA 932 EUR
MLDVR8222 MLDVR8223 MLDVR8330
Potassium assay kits, FLIPR®
Testen Cellulaire assays
The FLIPR® Potassium Assay Kit measures functional activity of ligand- and voltage-gated potassium channels. The homogeneous, no-wash assay protocol provides a large signal window and high Z' values.
  • Functional measurement of potassium channel activity in a cell-based assay
  • Homogenous no-wash protocol reduces well-to-well variation and simplifies the workflow
  • Expanded signal window compared to non-homogenous assay
  • Rapid procedure with less hands‐on time
The FLIPR® Potassium Assay Kit exploits the permeability of thallium ions (Tl+) through both voltage- and ligand-gated potassium (K+) channels. In this assay, a novel, highly-sensitive Tl+ indicator dye is utilized which produces a bright fluorescent signal upon the binding to Tl+ conducted through potassium channels. The intensity of the Tl+ signal is proportional to the number of potassium channels in the open state; therefore it provides a functional indication of the potassium channel activities. In addition, one of our proprietary masking dyes is employed to further reduce background fluorescence for improved signal/noise ratio.