Beakers, aluminium
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
39047.KT 39048.KT 39049.KT
39047.KTEA 97.3 EUR
39047.KT 39048.KT 39049.KT
Beakers, aluminium
Bekers
Beaker, aluminium, with pourout lip.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Aluminum is the most abundant metallic element on the earth and has many desirable physical and chemical properties. A highly impervious oxide film (approximately 5 nm thick) is resistant to corrosion by seawater, and other aqueous and chemical solutions. Additionally, this element offers chemical stability in the presence of most organic compounds. Aluminum's reactivity increases with temperature, therefore, the use ofMore Product Information