Dishes, aluminium, flat bottom, straight wall
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
39072.KH 39072.UD
39072.KHEA 38.4 EUR
39072.KH 39072.UD
Dishes, aluminium, flat bottom, straight wall
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Dishes, Aluminium, flat bottom, straight sides.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Aluminum is the most abundant metallic element on the earth and has many desirable physical and chemical properties. A highly impervious oxide film (approximately 5nm-thick) is resistant to corrosion by seawater, and other aqueous and chemical solutions. Additionally, this element offers chemical stability in the presence of most organic compounds. Aluminum's reactivity increases with temperature, therefore, the use of aluminum labware at high temperatures is not recommended.