Test sieves, US standard
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40181.NK 44753.NK 43657.NK 43659.NK
40181.NKEA 716 EUR
40181.NK 44753.NK 43657.NK 43659.NK
Test sieves, US standard
Zeven Testzeven
The brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments.
  • Superior quality guarantees accuracy and reproducibility
  • Sturdy construction ensures easy stacking
Sieve covers, made of polished brass, have a recessed wire handle.