Biopsy punches
Leverancier: LABONORD MISCELLANEOUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48301 48401 48501 48801 48601
LBMI48301EA 126 EUR
LBMI48301 LBMI48401 LBMI48501 LBMI48801 LBMI48601
Biopsy punches
Biopsie stoten
This biopsy punch makes it possible to perform skin biopsies in various locations with great precision and minimal tissue trauma. The stainless steel blade is ultra sharp thanks to high-precision micro-grinding technology.
  • Ribbed handle ensures a safe grip and optimal control during specimen collection
  • Size information on each punch
  • Sterile packed disposable punch makes grinding and sterilising unnecessary