Hand cleansing wipes, pre-saturated, Kresto® Special WIPES
Leverancier: Deb-STOKO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ULT150W ULT70W
115-2381EA 153.8 EUR
115-2381 115-2384
Hand cleansing wipes, pre-saturated, Kresto® Special WIPES
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Vochtige doekjes
Tough, extra large wipes impregnated with a mild ester solvent-based liquid formula for highly effective removal of paints, inks, resins, adhesives and most other print and paint shop contaminants. For use without water in industrial environments. Suitable for use on-site and for mobile workers.
  • No water needed: Impregnated with an advanced hi-tech formulation solvent liquid that includes dibasic esters
  • Wipes are strong and absorbent, acting quickly and effectively leaving no sticky feeling
  • Highly effective and environmentally friendly moisturiser for conditioning the skin
  • Perfumed with a pleasant citrus fragrance
  • Stored in a sealed container to protect from contamination
  • Silicone-free
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904