Regenerating hand and face cream, Stokolan® classic
Leverancier: Deb-STOKO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SCL100ML SCL1L
115-2156EA 60 EUR
115-2156 115-2343
Regenerating hand and face cream, Stokolan® classic
Huidverzorging Huidcrèmes Hand- en bodylotions
Stokolan® classic is a concentrated moisturising and conditioning cream that helps to restore moisture in normal and dry skin. Ideal for use in industrial and healthcare environments, especially where employees are prone to dry or stressed skin. Suitable for hands and face.
  • Helps work stressed skin to restore its natural resistance
  • With 9,5% glycerine and urea: Enhances skin elasticity and soothes the skin
  • Contains skin nourishing Lanolin of pharmaceutical quality: Restores natural oils and moisturises to prevent dry and rough skin
  • Dermatologically confirmed skin compatibility
  • Suitable for sensitive and eczema-prone skin
  • Silicone-free and paint compatible
Certificaten: ECARF Quality Seal for allergy friendliness
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904