Culture tubes, disposable, with screw thread finish, KIMBLE®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP73750-16100 SP73750-16125 SP73750-20150
SCERSP73750-16100EA 934 EUR
SCERSP73750-16100 SCERSP73750-16125 SCERSP73750-20150
Culture tubes, disposable, with screw thread finish, KIMBLE®
Buisjes Cultuurbuisjes
These culture tubes are manufactured from 51 expansion borosilicate glass. Suitable for tissue culture work and general bacteriological use.
  • Round-bottomed
  • Each tube has a chevron marking area
  • Modular trays are shrink-wrapped in plastic film
  • Supplied without caps
Certificaten: Glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.