Spinner flasks, adjustable hanging bar
Leverancier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1967-00025 1967-00100A 1967-00250 1967-00500 1967-01000 1967-03000
BELC1967-00025EA 185.65 EUR
BELC1967-00025 BELC1967-00100A BELC1967-00250 BELC1967-00500 BELC1967-01000 BELC1967-03000
Spinner flasks, adjustable hanging bar
Kolven Spinnerflessen
Spinner flasks, made of borosilicate glass,with PTFE impeller assembly.
  • Height adjustable impeller
  • Impeller assembly rides on glass bearing for friction free operation
  • Flat bottom
The hanging bar spinner flasks have black phenolic rubber-lined central and sidearm caps for 25 and 50 ml flasks.