Capillary columns, BP20 (Wax)
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
054405 054421 054422 054424 054425 054427 054428 054431 054433 054434 054436 054437 054440 054442 054443 054448 054450 054451 0544515 054456 054462 054415 054438 054439 054444 054445 054457 054458
568-0145EA 404 EUR
568-0145 558-0324 558-0325 568-0147 568-0148 558-0253 558-0327 568-0150 558-0328 568-0152 558-0329 568-0153 568-0156 568-0157 568-0158 568-0162 568-0163 558-0330 568-0164 568-0166 568-0169 558-0323 568-0154 568-0155 568-0159 568-0160 568-0167 568-0168
Capillary columns, BP20 (Wax)
Kolommen voor chromatografie
These polyethylene glycol columns are suitable for the analysis of alcohol, free acids, fatty acid methyl esters, aromatics, solvents, essential oils.
  • Polar phase
  • Cross-linked for stability and washing
Suitable replacement for: DB-Wax, HP-20M, Supelcowax 10, CB-Wax, Stabilwax, Carbowax, HP-Innowax, Rtx-WAX, PE-WAX, RH-WAX, ZB-WAX, TRWAX.
Max. temperature: 0,1 to 1,0 µm film thickness 20 °C to 260/280 °C; >1,0 µm film thickness 20 °C to 240/260 °C