Inline check valves, non-metallic ¹/₄-28 and 10-32, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CV-3320 CV-3321 CV-3324 CV-3325 CV-3330 CV-3340
554-3061EA 291 EUR
554-3061 554-3062 UPCHCV-3324 UPCHCV-3325 554-3063 554-3064
Inline check valves, non-metallic ¹/₄-28 and 10-32, Upchurch Scientific®
Ventielen Terugslagventielen
These biocompatible PEEK and perfluoroelastomer inline check valves are designed for applications where low flow path volume is critical, such as delivery to lab-on-a-chip, single-cell analysis and micro- or nano-LC post-column derivatisation.
  • Low cracking pressure of 1 psi (0,07 bar)
  • Excellent chemical resistance
  • Designed to provide excellent backflow protection for sensitive equipment
  • Perfect for use with corrosive fluids or biological samples