Inline cartridge check valves, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CV-3000 CV-3000NF CV-3001
554-3057EA 267 EUR
554-3057 554-3058 554-3059
Inline cartridge check valves, Upchurch Scientific®
Ventielen Terugslagventielen
Inline cartridge check valves are designed to limit flow in one direction and deliver a low cracking pressure. Ideal placement for these valves would be on the effluent side of the column to prevent post-column derivatisation agents from flowing back and poisoning the column if the HPLC mobile phase pump were to shut down.
  • Low cracking pressures
  • Designed to limit flow to one direction
  • Pressure rating up to 1000 psi (69 bar)