Lijnfilters, Polydisc™ GW, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10463400 10463401
514-0088EA 274 EUR
514-0088 514-0089
Lijnfilters, Polydisc™ GW, Whatman™
Filters Lijnfilters
Cytiva's Polydisc GW (Ground Water) is an in-line filter specifically designed for preparation of ground water samples for dissolved heavy metal analysis.
  • Hydrophilic, durable nylon membrane is compatible with aqueous samples
  • 100% quartz fiber prefilter with low heavy metal content allows filtration of larger volumes of even heavily particle-laden samples
  • Certified low background metal values for heavy metal testing
  • Large 20,4 cm² filtration area
  • Tubing nozzle compatible with 6 - 14 mm Int.Ø hose
Polydisc GW’s large filter surface, durable nylonMore Product Information