Lijnfilters, Polydisc™ HD, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6728-5050 6728-5100 2227
516-4464EA 173 EUR
516-4464 516-4465 WHAT2227
Lijnfilters, Polydisc™ HD, Whatman™
Filters Lijnfilters
Polydisc HD (Heavy Duty) from Cytiva is a durable all-polypropylene in-line filter for large volumes of liquid samples containing even harsh chemical solvents.
  • In-line, hose-connected filter with up to 1000 ml capacity
  • High flow filter with durable all-polypropylene construction
  • Broad aqueous and solvent compatibility
  • 99% liquid retention rating
  • Available in 5 and 10 µm pore size
  • Recyclable after use
Labs that regularly process large sample volumes with different chemical compositions, pH values, and temperatures require a filter that is fast, durable,More Product Information
Order Now
Specificaties
Environmentally Preferable