Lijnfilters, Polydisc™ TF, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6720-5001 6720-5002 6720-5045 6721-5010
516-1106EA 173 EUR
516-1106 514-0492 514-0493 514-0494
Lijnfilters, Polydisc™ TF, Whatman™
Filters Lijnfilters
Whatman Polydisc TF In-Line Filters from Cytiva are lightweight devices with high chemical and solvent resistance that are particulraly suitable for protective vents or in-line filtration applications.
  • Chemically resistant hydrophobic PTFE membrane is well suited as a solvent filter for many different solutions
  • Filter can be autoclaved multiple times without losing integrity
  • Integrity-testable (bubble point or water breakthrough pressure in situ methods)
  • Lightweight to prevent tube collapsing
Filtering organic solvents and aggressive solutions calls for a chemically resistant, durable membrane. Cytiva's Polydisc TF features a PTFE membrane resistant to such compounds. The 1 µm membrane features a polypropylene prefilter for solvent filtration containing high particulate density in order to extend the useful life of the membrane.
Connections: Tubing nozzle 6 to 10 mm ID hose