Crucibles, tall form
Leverancier: CHEMPUR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TI2/6 TI2/7 TI2/8 TI2/9 TI2/10
459-4006EA 0 EUR
459-4006 459-4007 459-4008 459-4009 459-4010
Crucibles, tall form
Kroezen
Platinum/iridium 90/10 crucibles with reinforced rim.