Schlenk tubes, KIMBLE®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP218710-0015 SP218710-0025 SP218710-0050 SP218710-0100
SCERSP218710-0015EA 259 EUR
SCERSP218710-0015 SCERSP218710-0025 SCERSP218710-0050 SCERSP218710-0100
Schlenk tubes, KIMBLE®
Spruitstukken
This Schlenk glass tube is designed for type 216050 manifolds and performs well under both thermal and pressure-type reactions.
Certificaten: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.