Verification standard kit, MultiDirect
Leverancier: Lovibond
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
215650
215670
LOVI215650EA
199.9
EUR
LOVI215650
LOVI215670
Verification standard kit, MultiDirect
Standaarden
Standaarden voor kleurreferentie en spectroscopie
Verification accuracy of photometers and validity of results.
- Check the performance of photometers over the full measurement range
- Results can be checked against tolerances provided on the certificate of conformity
Leveringsinformatie: Contains zero solution in sealed photometer vial, 6× verification standard solution in sealed photometer vial, light shield cap, certificate of conformity, instruction sheet.