Syringes for HPLC autosamplers, PerkinElmer instruments
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
80062 80262 81460 81262 81360 80962 81062 81162 81160
HAMI80062EA 186 EUR
HAMI80062 HAMI80262 HAMI81460 HAMI81262 HAMI81360 HAMI80962 HAMI81062 HAMI81162 HAMI81160
Syringes for HPLC autosamplers, PerkinElmer instruments
Spuiten Injectiespuiten voor chromatografie
Gastight syringes for use with gases or liquids. Designed with or without ChemSeal (C) termination.
  • PTFE plunger tip creates a leak-free seal
  • Syringe glass barrel and plunger are interchangeable
Cheminert (C or CX) - This termination is a 1/4"−28 fitting designed to fit directly into instrumentation. The X-style, meaning plunger stop, on this syringe is designed to limit wear and tear on the plunger tip.