ESD safety shoes, lace-up, uvex 1 x-tended support, 8514
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8514.2/35 8514.2/36 8514.2/37 8514.2/38 8514.2/39 8514.2/40 8514.2/41 8514.2/42 8514.2/43 8514.2/44 8514.2/45 8514.2/46 8514.2/47 8514.2/48 8514.2/49 8514.2/50 8514.2/51 8514.2/52
UVEX8514.2/35EA 116 EUR
UVEX8514.2/35 UVEX8514.2/36 UVEX8514.2/37 UVEX8514.2/38 UVEX8514.2/39 UVEX8514.2/40 UVEX8514.2/41 UVEX8514.2/42 UVEX8514.2/43 UVEX8514.2/44 UVEX8514.2/45 UVEX8514.2/46 UVEX8514.2/47 UVEX8514.2/48 UVEX8514.2/49 UVEX8514.2/50 UVEX8514.2/51 UVEX8514.2/52
ESD safety shoes, lace-up, uvex 1 x-tended support, 8514
Schoenen en laarzen
Metal-free, extremely lightweight, flexible, multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, plus a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Virtually seamless design eliminates potential pressure points and blisters
  • Elastic laces for quick, individual adjustment
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Flexible, metal-free, puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black
Width: 11
Certificaten: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1P, SRC