Accessories for syringes, SampleLock™
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35080 35083 35081
HAMI35080EA 45.6 EUR
HAMI35080 HAMI35083 HAMI35081
Accessories for syringes, SampleLock™
Spuiten Injectiespuiten voor chromatografie
These are a series of adapters that convert a large hub removable needle (RN termination) syringe to a Luer connection. There are three different removable adapters for 250 µl and larger syringes.
The adapters convert the RN termination into either a Male Luer, Male Luer Lock or a Female Luer Lock connection. The adapter has a port inner diameter of 0,99 mm that allows for sample spiking through a syringe fitted with a 26 gauge needle or smaller.