Containers, Click Pack, nestable, with screw lid, UN-marking
Leverancier: CURTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4420-60-004 4515-60-004 4525-70-004
CURT4420-60-004EA 25.2 EUR
CURT4420-60-004 CURT4515-60-004 CURT4525-70-004
Containers, Click Pack, nestable, with screw lid, UN-marking
Vaten, vaten, trommels
Nestable containers, made of white HDPE, with dark blue screw lid. For storage and shipment of powders and pastes.
  • Water vapour tight
  • Tamper evident
  • Stackable
  • UN marking (1H2) for hazardous solids
  • Smooth inside
  • Food grade
Easy to close and open with just a quarter-turn. Take up less storage space when empty and help reduce your logistic cost.
Certificaten: UN-X certified (1H2) for solids