Cupric citrate solution 0.1 M USP Test Solution (TS)
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Copper citrate, Copper(II) citrate, Cuprocitrol
85342.180EA 48.9 EUR
85342.180 85342.260
Cupric citrate solution 0.1 M USP Test Solution (TS)
Cupric citrate
Ready to use solutions.
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Formule: C₆H₄Cu₂O₇
Kookpunt: 100 °C
Dichtheid: 1,31293 g/cm³ (20 °C)
CAS nummer: 866-82-0
EINECS: 0
UN: 3082
ADR: 9,III