Antibiotic assay discs, multodisc
Leverancier: LIOFILCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
95240 95250 95260 95290 95230
LIOF95240EA 201 EUR
LIOF95240 LIOF95250 LIOF95260 LIOF95290 LIOF95230
Antibiotic assay discs, multodisc
Testsystemen voor microbiologie Antibioticatesten
Multodisc with eight antimicrobial agents on a ring.
  • Each specially for a microorganism
Amikacin (AK), Ampicillin (AMP), Ampicillin-Sulbactam (AMS), Carbenicillin (CAR), Cefoxitin (FOX), Ceftazidime (CAZ), Cefuroxime (CXM), Chloramphenicol (C), Clindamycin (CD), Erythromycin (E), Gentamicin (CN), Levofloxacin (LEV), Linezolid (LNZ), Metronidazole (MTZ), Netilmicin (NET), Nitrofurantoin (F), Norfloxacin (NOR), Penicillin G (P), Piperacillin-Tazobactam (TZP), Rifampicin (RD), Streptomycin (S), Teicoplanin (TEC), Tetracycline (TE), Ticarcillin (TC), Tobramycin (TOB), Trimethoprim-Sulfamethoxazole (SXT), Vancomycin (VA). The numbers shows the concentration in μg.