Spanner wrenches
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28-9647-78 28-9647-76 28-9647-77
28-9647-78EA 100 EUR
28-9647-78 28-9647-76 28-9647-77
Spanner wrenches
Moersleutels
These spanner wrenches are used with HiScale™ columns. To readjust the position of the adapter in the column a spanner can be used to turn the end cap. To prevent the column from rotating, a second spanner can be used to hold the end housing.