Filling and storage solution for combined/reference electrodes
Leverancier: Hach
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C20C500
C20C320
HACHC20C500EA
52
EUR
HACHC20C500
HACHC20C320
Filling and storage solution for combined/reference electrodes
Elektrolyten
Vuloplossingen voor elektrodes
Shelf life: 48 months from production date.