Conductivity standards, KCL
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C20C270 C20C250
HACHC20C270EA 53.5 EUR
HACHC20C270 HACHC20C250
Conductivity standards, KCL
Standaarden Geleidbaarheidsstandaarden
The potassium chloride standard solutions are used for calibrating conductivity cells.