Luminescent bacteria test kit
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LCK480 LCK481 LCK482 LCK484 LCK487 LCK488 LCK491
HACHLCK480EA 481 EUR
HACHLCK480 HACHLCK481 HACHLCK482 HACHLCK484 HACHLCK487 HACHLCK488 HACHLCK491
Luminescent bacteria test kit
Testsystemen voor microbiologie Micro-organisme testen
The luminescent bacteria test is a simple operation to carry out as a chemical analysis. It provides quick results within maximum test time of 30 min.
  • Long-life preserved bacteria
  • Viability indicated by natural luminescence
  • Effortless compensation for interferences
The preserved luminescent bacteria are reactivated before the test is started. Toxicity is indicated by the luminescence, more the sample inhibits luminescence, more toxic it is.
Certificaten: Conforms to international EN ISO 11348 standard.