Xyleen, EMSURE® ISO, ACS, Reag. Ph. Eur. for analysis
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Dimethylbenzene
M-Clarity™ quality level = MQ300
1.08297.2500 1.08297.4000
1.08297.2500EA 233 EUR
1.08297.2500 1.08297.4000
Xyleen, EMSURE® ISO, ACS, Reag. Ph. Eur. for analysis
Xyleen
1.08297 Xylene (isomeric mixture) EMSURE® is a real isomeric mixture of ortho-, meta- and para-Xylene.
Formule: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Kookpunt: 140
Smeltpunt: –25 °C
Dichtheid: 0.86…0.88 g/cm³ (20 °C)
Vlampunt: 25
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00077264
CAS nummer: 1330-20-7
UN: 1307
ADR: 3 III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM