Normal phase HPLC columns, Luna® Silica(2)
Leverancier: PHENOMENEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
00F-4162-B0 00G-4274-E0
PHEN00F-4162-B0EA 0 EUR
PHEN00F-4162-B0 PHEN00G-4274-E0
Normal phase HPLC columns, Luna® Silica(2)
Kolommen voor chromatografie
Luna® Silica(2) performance is of ultrapure metal-free silica (99,99% purity) and meticulous care is given to the quality control of surface smoothness, pore structure and pore consistency to ensure particles of uniform structure and enhanced mechanical strength.
  • Low percentage of 'fines' from damaged silica leading to lower backpressures and enhanced column performance and lifetimes
  • High column bed stability enhanced by particle shape uniformity
Commonly applied for Alprazolam, Hydrocodone bitartrate, Hydrocortisone, Fat Soluble Vitamins, Phthalates, Fatty Acids, Lutein, Lycopene, Estradiol.