Capillary columns, HP-chiral β
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19091G-B133 19091G-B213 19091G-B233 19091G-B233E
HEWL19091G-B133EA 826 EUR
HEWL19091G-B133 HEWL19091G-B213 HEWL19091G-B233 HEWL19091G-B233E
Capillary columns, HP-chiral β
Kolommen voor chromatografie
Phenyl-based polymer provides low bleed and does not interfere with nitrogen-specific detectors.
  • Beta-cyclodextrin in (35%-Phenyl)-methylpolysiloxane
  • Chiral separations without chiral specific derivatisation
  • Available in two concentrations of beta-cyclodextrin: 10% and 20%
  • 20% beta-cyclodextrin best choice for initial screening