Safety dispensing cans
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
16768 16769 16770
MSGA16768EA 225 EUR
MSGA16768 MSGA16769 MSGA16770
Safety dispensing cans
Veiligheidscontainers
Safety cans, made of high quality and chemically resistant stainless steel with PTFE gaskets. For safe and comfortable handling of small amounts of flammable liquids.
  • Easy handling, ergonomic design and high stability
  • Flame traps and explosion-proof
  • Pressure relief valve (vent at about 0,3 bar)
  • Self-closing metering devices
  • Moisteners for safe cleaning with solvents