GC columns, HayeSep R
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G3591-80100 G3591-80124
HEWLG3591-80100EA 0 EUR
HEWLG3591-80100 HEWLG3591-80124
GC columns, HayeSep R
Kolommen voor chromatografie
Packed GC columns are designed and manufactured to offer excellent and reproducible performance for all sample types associated with packed column separations, most important in the hydrocarbon processing industry.
  • Highly efficient and rigorous packing technology assures column-to-column reproducibility and ultimate efficiency
  • UltiMetal treated stainless steel tubing allows for improved inertness and peak shape performance