Peptide mapping columns, AdvanceBio
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
653950-902 653950-302 653750-902 651750-902 655750-902 853750-911 851725-911 850750-911 863600-911
HEWL653950-902EA 1091 EUR
HEWL653950-902 HEWL653950-302 HEWL653750-902 HEWL651750-902 HEWL655750-902 HEWL853750-911 HEWL851725-911 HEWL850750-911 HEWL863600-911
Peptide mapping columns, AdvanceBio
Kolommen voor chromatografie
These peptide mapping columns are designed to provide high- resolution peptide maps for protein identification and determination of post translation modifications. These columns provide the ability to quickly resolve and identify amino acid substitutions/modifications in a protein primary sequence.
  • Greater analytical confidence
  • Increased flexibility
These columns delivers consistent, exceptional performance for the separation and characterisation of peptides and proteins, antibodies, conjugates, new biological entities, and biopharmaceuticals. These advanced biocolumns are superficially porous and have a 120 Å pore size which is optimum for the analysis of the peptides produced by enzymatic digest of proteins. The 2,7 µm particle size enables UHPLC performance on HPLC systems, which delivers high resolution separations 2 to 3 times faster than with fully porous HPLC columns.