GHOST ESD Ladies Safety Shoes, Low, S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
74171/35 74171/36 74171/37 74171/38 74171/39 74171/40 74171/41 74171/42
114-7333EA 81.9 EUR
114-7333 114-7334 114-7335 114-7336 114-7337 114-7338 114-7339 114-7340
GHOST ESD Ladies Safety Shoes, Low, S3
Schoenen en laarzen
ESD ladies safety shoe, GHOST, available in white finish.
  • Hydrophobised microfibre
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole ESD PRO Lady grey
  • MONO-PU sole MISS L10
  • Steel toe cap
Ladies shoes feature a non-metallic, penetration-resistant midsole.
Certificaten: EN ISO 20345 S3 SRC, form A.