Zebron™ ZB-WAX GC Columns
Leverancier: PHENOMENEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7KG-G007-11 7KK-G007-22
PHEN7KG-G007-11EA 0 EUR
PHEN7KG-G007-11 PHEN7KK-G007-22
Zebron™ ZB-WAX GC Columns
Kolommen voor chromatografie
High polarity column with low bleed (MS certified) for separating polar complex mixtures. Recommended for alcohols, aldehydes, aromatics, basic compounds, essential oils, flavors and fragrances, glycols, pharmaceuticals, solvents, styrene and xylene isomers.
  • Bonded, solvent rinsable phase excellent for separating polar complex mixtures
  • GC/MS certified low bleed levels
  • Low activity for amines
  • Excellent separations of polar complex mixtures; widely used for profiling and fingerprinting
Temperature limits: 40 to 250/260 °C (Isothermal/TPGC).