Crucibles, aluminium
Leverancier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
00027331 00026763 51119870
METN00027331EA 501 EUR
METN00027331 METN00026763 METN51119870
Crucibles, aluminium
Kroezen
Aluminum is the standard DSC crucible material up to 640 °C. For solids, liquids and powder samples and excellent DSC performance.
  • Eliminates interaction between the sample and the crucible
  • High thermal conductivity and form stability
  • Ensures perfect thermal contact and no artifacts due to its flatness