Cleanroom Overalls Tyvek® IsoClean® model IC183B DS
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IC183BWHDS/M IC183BWHDS/L IC183BWHDS/XL IC183BWHDS/2X IC183BWHDS/3X IC183BWHDS/S IC183BWHDS/XS
113-8629EA 439 EUR
113-8629 113-8630 113-8631 113-8632 113-8633 113-8628 DUPOIC183BWHDS/XS
Cleanroom Overalls Tyvek® IsoClean® model IC183B DS
Wegwerpkleding
These unhooded overalls are made of Tyvek®, a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
  • Ultra-low linting fabric
  • Unhooded overall with bound neck
  • Tyvek® covered elasticated thumb loops
  • Tunnelled elastication at wrists and ankles; stitched-in elasticated waist
  • Front zipper closure with storm flap
  • Optimised shape and fit allowing full freedom of movement
  • Clean-processed and gamma-sterilised
  • Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
Garments made of Tyvek® IsoClean are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other critical or controlled environments.
Colour: White
Certificaten: Type 5B/6B, EN 1073-2 (Class 2), EN 14126, Cat. III
Verpakking: Double bagged. Packed in an ISO Class 4 certified cleanroom .