Cleanroom boot covers, Tyvek® IsoClean®, model IC458B 00, non sterile
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IC458BWH00/L IC458BWH00/M
DUPOIC458BWH00/LEA 283 EUR
DUPOIC458BWH00/L DUPOIC458BWH00/M
Cleanroom boot covers, Tyvek® IsoClean®, model IC458B 00, non sterile
Wegwerpkleding
These non sterile overboots are made of Tyvek®. This fabric is composed of flash spun high density polyethylene, providing an ideal balance of protection, durability, and comfort.
  • Ultra-low-linting fabric
  • Fixation ties
  • Ankle ties for secure fit
  • Gripper™ soles provide enhanced skid-resistance and durability
  • Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1µm in size). Ideal for use in biotech, pharmaceutical, medical device manufacturing, and electronics industries, as well as in other critical or controlled environments.
Certificaten: (EU) 2016/425; Cat III, Type PB [6-B]; EN 14126
Verpakking: Bulk packaged in double transparent poly liners. 100 per case.