Reversed phase HPLC columns, Synergi™ Max-RP
Leverancier: PHENOMENEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
00G-4337-E0 00F-4337-E0 00F-4337-B0 00D-4337-B0
PHEN00G-4337-E0EA 0 EUR
PHEN00G-4337-E0 PHEN00F-4337-E0 PHEN00F-4337-B0 PHEN00D-4337-B0
Reversed phase HPLC columns, Synergi™ Max-RP
Kolommen voor chromatografie
Synergi™ Max-RP columns have a densely bonded C12 phase providing an alternative hydrophobic selectivity to traditional C18 columns.
  • Hydrophobic retention similar to a C18 with improved results
  • Sharper peaks for basic and tailing compounds
  • Ideal for the separation of basic compounds under neutral pH