Electric lab burner with regulator
Leverancier: HORO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HORO7001-7002-7003EA 0 EUR
HORO7001-7002-7003
Electric lab burner with regulator
Verwarmers Elektrische branders
Electric laboratory burner which can be operated as a 'stand alone' burner or with a glass support and/or power regulator.
  • Clean and odourless operation in regular atmosphere
  • No carbon black and no chemical reduction of the flame
  • Homogeneous heating without peaks or overheating
  • No risk of flames, explosions or danger due toxic gas
Base Ø: 125 mm
Height: 170 mm