Wafer tweezers, ESD-safe
Leverancier: OUTILS RUBIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43LB-4POLY 43LB-8POLY
OUTI43LB-4POLYEA 43.8 EUR
OUTI43LB-4POLY OUTI43LB-8POLY
Wafer tweezers, ESD-safe
Pincetten
Polyamide. These cleanroom compatible tweezers are specially designed to handle different fragile and delicate materials of gallium or silicone wafers.
  • Graduated lower paddles prevent the wafer from slipping
  • ESD safe (103Ω x cm)
  • Heat resistance up to 175 °C (short exposure 300 °C)