Visual comparator test kits, chlorine
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
223101 223102 223103 1454200 2129000
705-0102EA 144 EUR
705-0102 705-0103 HACH223103 705-0069 705-0099
Visual comparator test kits, chlorine
Milieutestkits
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case for 100 tests
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Leveringsinformatie: Elke testkit bevat alle benodigde reagentia, reactiecuvetten, een kleurvergelijkingskaart en uitgebreide gebruiksinstructies.