Powder pillows, chromium
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1271099 1206699 204399 204499
HACH1271099EA 53.6 EUR
HACH1271099 HACH1206699 HACH204399 HACH204499
Powder pillows, chromium
Milieutestkits
ChromaVer 3 reagent for hexavalent chromium determination by the Diphenylcarbohydrazide Method. USEPA accepted for reporting for wastewater analyses using Hach Method 8023.